3-Chloro-2,6-xylidine hydrochloride 99%

Warning

Synonyms: 3-Chloro-2,6-dimethylaniline hydrochloride

B21851-06 B21851-14 B21851-14 B21851-06
AAB21851-14EA 88.76 USD
CAAAB21851-14 AAB21851-14 AAB21851-06 CAAAB21851-06
3-Chloro-2,6-xylidine hydrochloride 99%
3-Chloro-2,6-xylidine hydrochloride
Formula: C₈H₁₁Cl₂N
MW: 192.09
Melting Pt: ca 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02683075
CAS Number: 72725-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR