n-Octyl-L-2-pyrrolidinone-5-carboxylate 98%

B21838-14 B21838-06 B21838-06 B21838-14
AAB21838-14EA 215.77 USD
CAAAB21838-14 AAB21838-14 AAB21838-06 CAAAB21838-06
n-Octyl-L-2-pyrrolidinone-5-carboxylate 98%
n-Octyl-L-2-pyrrolidinone-5-carboxylate
Formula: C₁₃H₂₃NO₃
MW: 241.33 g/mol
Boiling Pt: 140 °C/0.06 mm
Storage Temperature: Ambient
MDL Number: MFCD00236053
CAS Number: 4931-70-8
EINECS: 225-569-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR