2,2,4-Trimethyl-1,3-dioxolane 99%

Supplier: Thermo Scientific Chemicals
Warning

B21823-22 B21823-36 B21823-14 B21823-22 B21823-36 B21823-14
AAB21823-36EA 491.99 USD
CAAAB21823-22 AAB21823-36 AAB21823-14 AAB21823-22 CAAAB21823-36 CAAAB21823-14
2,2,4-Trimethyl-1,3-dioxolane 99%
2,2,4-Trimethyl-1,3-dioxolane
Formula: C₆H₁₂O₂
Boiling Pt: 98 °C
Density: 0.897
Flash Pt: 32 °C(90 °F)
Storage Temperature: Ambient
MDL Number: MFCD00090841
CAS Number: 1193-11-9
EINECS: 214-766-0
UN: 3271
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR