2-Chlorobutyramide 98%

Supplier: Thermo Scientific Chemicals
Warning

B21783-06 B21783-03 B21783-03 B21783-06
AAB21783-03EA 28.86 USD
AAB21783-03 CAAAB21783-06 CAAAB21783-03 AAB21783-06
2-Chlorobutyramide 98%
2-Chlorobutyramide
Formula: C₄H₈ClNO
MW: 121.57 g/mol
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00041477
CAS Number: 7462-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR