2,5-Dimethylbenzoxazole 99%

Warning

B21780-03 B21780-03 B21780-06 B21780-06
AAB21780-06EA 81.27 USD
CAAAB21780-06 CAAAB21780-03 AAB21780-06 AAB21780-03
2,5-Dimethylbenzoxazole 99%
2,5-Dimethylbenzoxazole
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 218…219 °C
Density: 1,090
Storage Temperature: Ambient
MDL Number: MFCD00005773
CAS Number: 5676-58-4
EINECS: 227-136-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR