1,3-Propanedisulfonic acid ≥70% (w/v) in aqueous solution

Danger

B21776-22 B21776-22 B21776-14 B21776-14
AAB21776-14EA 140.14 USD
CAAAB21776-22 CAAAB21776-14 AAB21776-14 AAB21776-22
1,3-Propanedisulfonic acid ≥70% (w/v) in aqueous solution
1,3-Propanedisulfonic acid
Formula: C₃H₈O₆S₂
MW: 204.21
Density: 1.33
Storage Temperature: Ambient
MDL Number: MFCD00191482
CAS Number: 21668-77-9
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR