Ethyl-3-(2-furfurylthio)propionate 97%

Synonyms: Ethyl 3-(furfurylthio)propionate

B21766-18 B21766-09 B21766-09 B21766-18
AAB21766-09EA 51.73 USD
AAB21766-09 CAAAB21766-09 AAB21766-18 CAAAB21766-18
Ethyl-3-(2-furfurylthio)propionate 97%
Ethyl 3-(2-furfurylthio)propionate
Formula: C₁₀H₁₄O₃S
Boiling Pt: 120…122 °C/0.5 mm
Density: 1.130
Flash Pt: 99 °C(210 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051819
CAS Number: 94278-27-0
EINECS: 304-716-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR