Isobutyramide 99%

Supplier: Thermo Scientific Chemicals
Warning

B21641-36 B21641-36 B21641-14 B21641-14 B21641-22 B21641-22
AAB21641-22EA 185.4 USD
CAAAB21641-36 CAAAB21641-14 AAB21641-22 CAAAB21641-22 AAB21641-36 AAB21641-14
Isobutyramide 99%
Isobutyramide
Formula: C₄H₉NO
MW: 87.12 g/mol
Boiling Pt: 216…220 °C
Melting Pt: 127…129 °C
Density: 1,013
Storage Temperature: Ambient
MDL Number: MFCD00008019
CAS Number: 563-83-7
EINECS: 209-265-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR