2,2,4,4-Tetrafluoro-1,3-dithietane 97%

Warning

B21620-18 B21620-09 B21620-09 B21620-18
AAB21620-18EA 111.88 USD
AAB21620-18 AAB21620-09 CAAAB21620-18 CAAAB21620-09
2,2,4,4-Tetrafluoro-1,3-dithietane 97%
2,2,4,4-Tetrafluoro-1,3-dithietane
Formula: C₂F₄S₂
Boiling Pt: 47…49 °C
Density: 1.60
Flash Pt: 36 °C(96 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053538
CAS Number: 1717-50-6
EINECS: 217-001-9
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR