1-Chloro-2-cycloheptylethane 96%

Warning

B21604-14 B21604-14 B21604-06 B21604-06
AAB21604-06EA 58.6 USD
AAB21604-06 CAAAB21604-06 CAAAB21604-14 AAB21604-14
1-Chloro-2-cycloheptylethane 96%
1-Chloro-2-cycloheptylethane
Formula: C₉H₁₇Cl
Storage Temperature: Ambient
MDL Number: MFCD01632144
CAS Number: 84226-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR