2-Cyanobenzamide 98%

Warning

B21596-06 B21596-03 B21596-03
AAB21596-06EA 407.31 USD
AAB21596-06 AAB21596-03 CAAAB21596-03
2-Cyanobenzamide 98%
2-Cyanobenzamide
Formula: C₈H₆N₂O
Melting Pt: 172…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00014797
CAS Number: 17174-98-0
EINECS: 241-224-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR