3-Amino-4-bromo-1-methyl-1H-pyrazole 97+%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 4-Bromo-1-methyl-1H-pyrazol-3-amine

B21592-01 B21592-03 B21592-03 B21592-01
AAB21592-03EA 107 USD
AAB21592-03 AAB21592-01 CAAAB21592-01 CAAAB21592-03
3-Amino-4-bromo-1-methyl-1H-pyrazole 97+%
3-Amino-4-bromo-1-methyl-1H-pyrazole
Formula: C₄H₆BrN₃
MW: 176.02 g/mol
Melting Pt: 96…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00663065
CAS Number: 146941-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR