4'-Isopropylacetanilide 97+%

Supplier: Thermo Scientific Chemicals

Warning

B21591-14 B21591-14 B21591-06 B21591-06
AAB21591-06EA 40.3 USD
CAAAB21591-06 AAB21591-06 AAB21591-14 CAAAB21591-14
4'-Isopropylacetanilide 97+%
4'-Isopropylacetanilide
Formula: C₁₁H₁₅NO
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00026406
CAS Number: 5702-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR