1,4-Diisopropylcyclohexane (mixture of cis and trans isomeres) 98%

Danger

B21587-14 B21587-14 B21587-06
AAB21587-14EA 196.47 USD
CAAAB21587-14 AAB21587-14 CAAAB21587-06
1,4-Diisopropylcyclohexane (mixture of cis and trans isomeres) 98%
1,4-Diisopropylcyclohexane (mixture of cis- and trans-isomeres)
Formula: C₁₂H₂₄
Boiling Pt: 214…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00019366
CAS Number: 22907-72-8
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR