3-(Trifluoromethoxy)cinnamic acid 96%

Warning

Synonyms: 3-(3-(Trifluoromethoxy)phenyl)acrylic acid

B21560-03 B21560-03 B21560-01 B21560-01
AAB21560-01EA 64.86 USD
AAB21560-01 CAAAB21560-03 AAB21560-03 CAAAB21560-01
3-(Trifluoromethoxy)cinnamic acid 96%
3-(Trifluoromethoxy)cinnamic acid
Formula: C₁₀H₇F₃O₃
MW: 232.16 g/mol
Melting Pt: 92…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00066337
CAS Number: 168833-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR