10-Chloro-3-decyne 98%

B21556-14 B21556-06 B21556-06 B21556-14
AAB21556-06EA 54.6 USD
CAAAB21556-06 AAB21556-06 AAB21556-14 CAAAB21556-14
10-Chloro-3-decyne 98%
10-Chloro-3-decyne
Formula: C₁₀H₁₇Cl
Boiling Pt: 122 °C/20 mm
Density: 0.92
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01321154
CAS Number: 18295-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR