trans-4-tert-Pentylcyclohexanol 97%

Warning

Synonyms: trans-4-tert-Amylcyclohexanol

B21479-06 B21479-06 B21479-03 B21479-03
AAB21479-06EA 168.72 USD
CAAAB21479-03 AAB21479-06 CAAAB21479-06 AAB21479-03
trans-4-tert-Pentylcyclohexanol 97%
trans-4-tert-Pentylcyclohexanol
Formula: C₇H₁₄O
MW: 170.30 g/mol
Boiling Pt: 154…156 °C/40 mm
Melting Pt: 24…27 °C
Storage Temperature: Ambient
MDL Number: MFCD00067157
CAS Number: 20698-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR