4,6-Dichloro-2-(methylthio)pyrimidine 98%

Supplier: Thermo Scientific Chemicals

Danger

B21416-14 B21416-06 B21416-14 B21416-06
AAB21416-06EA 75.6 USD
AAB21416-06 AAB21416-14 CAAAB21416-06 CAAAB21416-14
4,6-Dichloro-2-(methylthio)pyrimidine 98%
4,6-Dichloro-2-(methylthio)pyrimidine
Formula: C₅H₄Cl₂N₂S
MW: 195.07 g/mol
Boiling Pt: 135…136 °C/14 mm
Melting Pt: 38…42 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006086
CAS Number: 6299-25-8
EINECS: 228-577-6
UN: 3263
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR