4,4'-Diacetoxybiphenyl 97%
B21403-14
B21403-06
B21403-03
B21403-06
B21403-03
B21403-14
AAB21403-14EA
288.6
USD
CAAAB21403-03
CAAAB21403-14
AAB21403-14
CAAAB21403-06
AAB21403-03
AAB21403-06
4,4'-Diacetoxybiphenyl 97%
4,4'-Diacetoxybiphenyl
Formula:
C₁₆H₁₄O₄ Melting Pt: 162…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035973 CAS Number: 32604-29-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...