1,3-Diisopropylcyclohexane (mixture of cis and trans isomeres) 97+%

Danger

B21390-06 B21390-14 B21390-14 B21390-06
AAB21390-06EA 60.75 USD
CAAAB21390-06 AAB21390-06 CAAAB21390-14 AAB21390-14
1,3-Diisopropylcyclohexane (mixture of cis and trans isomeres) 97+%
1,3-Diisopropylcyclohexane (mixture of cis- and trans-isomeres)
Formula: C₁₂H₂₄
Boiling Pt: 205…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00019365
CAS Number: 7045-70-7
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR