1,4,6,7-Tetramethylnaphthalene 98%

Warning

B21351-03 B21351-03 B21351-06 B21351-06
AAB21351-03EA 77.15 USD
CAAAB21351-06 CAAAB21351-03 AAB21351-03 AAB21351-06
1,4,6,7-Tetramethylnaphthalene 98%
1,4,6,7-Tetramethylnaphthalene
Formula: C₁₄H₁₆
Melting Pt: 63…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00049173
CAS Number: 13764-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR