N-Propyl-2-methylvalerate 98%

Warning

B21325-22 B21325-22 B21325-14 B21325-14
AAB21325-22EA 60.98 USD
AAB21325-22 CAAAB21325-14 AAB21325-14 CAAAB21325-22
N-Propyl-2-methylvalerate 98%
N-Propyl-2-methylvalerate
Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Flash Pt: 51 °C(124 °F)
Storage Temperature: Ambient
MDL Number: MFCD01632135
CAS Number: 6639-14-1
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR