3-(3-(Trifluoromethoxy)phenyl)propionic acid 95%
|
Warning
|
B21233-03
B21233-06
B21233-03
B21233-01
B21233-06
B21233-01
AAB21233-06EA
855.81
USD
CAAAB21233-06
AAB21233-06
CAAAB21233-03
CAAAB21233-01
AAB21233-01
AAB21233-03
3-(3-(Trifluoromethoxy)phenyl)propionic acid 95%
3-(3-(Trifluoromethoxy)phenyl)propionic acid
Formula:
C₁₀H₉F₃O₃ MW: 234.17 Storage Temperature: Ambient |
MDL Number:
MFCD01317834 CAS Number: 168833-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...