α-D(+)-Talose 97%
Supplier: Thermo Scientific Chemicals
B21226-77
B21226-02
B21226-77
B21226-02
AAB21226-02EA
376.99
USD
CAAAB21226-77
CAAAB21226-02
AAB21226-02
AAB21226-77
α-D(+)-Talose 97%
α-D-Talose
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Melting Pt: 133…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064906 CAS Number: 2595-98-4 EINECS: 219-996-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...