Diethyl (hydroxymethyl)phosphonate tech. 85%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21215-14
B21215-14
B21215-06
B21215-06
AAAB21215-14EA
122.99
USD
CAAAAB21215-14
AAAB21215-14
CAAAAB21215-06
AAAB21215-06
Diethyl (hydroxymethyl)phosphonate tech. 85%, Technical Grade
Diethyl (hydroxymethyl)phosphonate
Formula:
C₅H₁₃O₄P MW: 168.13 g/mol Boiling Pt: 124…126 °C/3 mm Density: 1.14 Flash Pt: 110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014418 EINECS: 221-391-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...