Cinnamyl propionate 97%

Warning

Synonyms: Propionic Acid Cinnamyl Ester

B21142-14 B21142-22 B21142-22 B21142-14
AAB21142-22EA 155.4 USD
AAB21142-22 CAAAB21142-14 AAB21142-14 CAAAB21142-22
Cinnamyl propionate 97%
Cinnamyl propionate
Formula: C₁₂H₁₄O₂
MW: 190.24 g/mol
Density: 1.04
Flash Pt: 100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048610
CAS Number: 103-56-0
EINECS: 203-124-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR