D-Glucose diethyl mercaptal 98%

Synonyms: D-Glucose Diethyl Dithioacetal

B21073-06 B21073-03 B21073-06 B21073-03
AAB21073-06EA 286.18 USD
CAAAB21073-03 AAB21073-06 AAB21073-03 CAAAB21073-06
D-Glucose diethyl mercaptal 98%
D-Glucose diethyl mercaptal
Formula: C₁₀H₂₂O₅S₂
MW: 286.40 g/mol
Melting Pt: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00004706
CAS Number: 1941-52-2
EINECS: 217-728-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR