5-Chloro-2-adamantanone ≥90%

Synonyms: 5-Chloroadamantan-2-one

B21028-03 B21028-03 B21028-06 B21028-06
AAB21028-03EA 80.23 USD
AAB21028-03 CAAAB21028-06 AAB21028-06 CAAAB21028-03
5-Chloro-2-adamantanone ≥90%
5-Chloro-2-adamantanone
Formula: C₁₀H₁₃ClO
MW: 184.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00798599
CAS Number: 20098-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR