2,5-Bis(4-nitrophenyl)-1,3,4-oxadiazole 97%

Warning

B20961-06 B20961-14 B20961-14 B20961-06
AAB20961-14EA 223.11 USD
CAAAB20961-14 AAB20961-14 CAAAB20961-06 AAB20961-06
2,5-Bis(4-nitrophenyl)-1,3,4-oxadiazole 97%
2,5-Bis(4-nitrophenyl)-1,3,4-oxadiazole
Formula: C₁₄H₈N₄O₅
Melting Pt: 310…312 °C
Storage Temperature: Ambient
MDL Number: MFCD00173659
CAS Number: 1044-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR