1-(5-Methyl-1-phenyl-1H-pyrazol-4-yl)-1-ethanone 97%

Warning

B20806-03 B20806-03 B20806-06 B20806-06
AAB20806-06EA 128.71 USD
CAAAB20806-03 CAAAB20806-06 AAB20806-06 AAB20806-03
1-(5-Methyl-1-phenyl-1H-pyrazol-4-yl)-1-ethanone 97%
1-(5-Methyl-1-phenyl-1H-pyrazol-4-yl)-1-ethanone
Formula: C₁₂H₁₂N₂O
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00020733
CAS Number: 6123-63-3
EINECS: 228-091-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR