2'-Fluoro-5'-(trifluoromethyl)acetophenone 97+%

Warning

B20777-03 B20777-03 B20777-06 B20777-06
AAB20777-06EA 233.1 USD
AAB20777-06 AAB20777-03 CAAAB20777-03 CAAAB20777-06
2'-Fluoro-5'-(trifluoromethyl)acetophenone 97+%
2'-Fluoro-5'-(trifluoromethyl)acetophenone
Formula: C₉H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD00061248
CAS Number: 202664-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR