3-Ethoxysalicylaldehyde 97%
|
Warning
|
B20702-30
B20702-18
B20702-18
B20702-30
AAB20702-30EA
134
USD
CAAAB20702-18
AAB20702-30
AAB20702-18
CAAAB20702-30
3-Ethoxysalicylaldehyde 97%
3-Ethoxysalicylaldehyde
Formula:
C₉H₁₀O₃ Boiling Pt: 263…264 °C Melting Pt: 66…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003323 CAS Number: 492-88-6 EINECS: 207-765-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...