Methyl-3,5-dibromobenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 3,5-dibromobenzoate
B20668-06
B20668-03
B20668-06
B20668-03
AAB20668-06EA
227.99
USD
AAB20668-06
AAB20668-03
CAAAB20668-03
CAAAB20668-06
Methyl-3,5-dibromobenzoate 98%
Methyl 3,5-dibromobenzoate
Formula:
C₈H₆Br₂O₂ MW: 293.94 g/mol Melting Pt: 61…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082648 CAS Number: 51329-15-8 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...