1,4-Benzodioxane-2-carboxylic acid 97%

Warning

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxine-2-carboxylic acid

B20661-03 B20661-06 B20661-03 B20661-06
AAB20661-06EA 128.76 USD
AAB20661-06 CAAAB20661-06 AAB20661-03 CAAAB20661-03
1,4-Benzodioxane-2-carboxylic acid 97%
1,4-Benzodioxane-2-carboxylic acid
Formula: C₉H₈O₄
MW: 180.16 g/mol
Melting Pt: 122…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00084888
CAS Number: 3663-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR