4-Methyl-4-hepten-3-ol 97%

Warning

B20658-03 B20658-06 B20658-14 B20658-06 B20658-03 B20658-14
AAB20658-06EA 96.28 USD
AAB20658-06 CAAAB20658-03 CAAAB20658-14 AAB20658-14 AAB20658-03 CAAAB20658-06
4-Methyl-4-hepten-3-ol 97%
4-Methyl-4-hepten-3-ol
Formula: C₈H₁₆O
Flash Pt: 59 °C(138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00046670
CAS Number: 81280-12-8
UN: 1987
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR