3-Ethyl-2-methyl-1-pentene 99%

Supplier: Thermo Scientific Chemicals
Danger

B20624-06 B20624-14 B20624-06 B20624-14
AAB20624-14EA 271.95 USD
CAAAB20624-06 AAB20624-14 AAB20624-06 CAAAB20624-14
3-Ethyl-2-methyl-1-pentene 99%
3-Ethyl-2-methyl-1-pentene
Formula: C₈H₁₆
MW: 112.22 g/mol
Density: 0.72
Storage Temperature: Ambient
MDL Number: MFCD00048664
CAS Number: 19780-66-6
UN: 3295
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR