2-Methyl-1-phenyl-1-pentanol erythro + threo 97%

B20596-06 B20596-14 B20596-06 B20596-14 B20596-03 B20596-03
AAB20596-14EA 344.1 USD
AAB20596-14 AAB20596-03 CAAAB20596-14 CAAAB20596-03 CAAAB20596-06 AAB20596-06
2-Methyl-1-phenyl-1-pentanol erythro + threo 97%
2-Methyl-1-phenyl-1-pentanol
Formula: C₁₂H₁₈O
MW: 178.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049211
CAS Number: 73177-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR