trans-2,4-Dimethoxy-β-nitrostyrene 99%

Warning

Synonyms: 2,4-Dimethoxy-β-nitrostyrene

B20564-06 B20564-03 B20564-03 B20564-06
AAB20564-06EA 80.93 USD
AAB20564-06 CAAAB20564-03 CAAAB20564-06 AAB20564-03
trans-2,4-Dimethoxy-β-nitrostyrene 99%
trans-2,4-Dimethoxy-β-nitrostyrene
Formula: C₁₀H₁₁NO₄
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00015922
CAS Number: 1891-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR