3-Chloroadamantane-1-carboxylic acid 97%

Warning

B20543-03 B20543-03 B20543-06 B20543-06
AAB20543-06EA 334.25 USD
CAAAB20543-03 AAB20543-06 AAB20543-03 CAAAB20543-06
3-Chloroadamantane-1-carboxylic acid 97%
3-Chloroadamantane-1-carboxylic acid
Formula: C₁₁H₁₅ClO₂
Storage Temperature: Ambient
MDL Number: MFCD00167831
CAS Number: 34859-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR