Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-3,5-dihydroxybenzamide 97%

Supplier: Alfa Aesar
Warning

B20493-18 B20493-09 B20493-18 B20493-09
AAB20493-09EA 55.83 USD InStock
CAAAB20493-18 AAB20493-09 AAB20493-18 CAAAB20493-09
4-Bromo-3,5-dihydroxybenzamide 97%
4-Bromo-3,5-dihydroxybenzamide


Formula: C₇H₆BrNO₃
MW: 232,03
Storage Temperature: Ambient
MDL Number: MFCD00017131
CAS Number: 13429-12-4
EINECS: 236-548-4
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty