2,4,4,6-Tetramethyl-1-oxa-3-aza-2-cyclohexene 95%

Warning

B20438-14 B20438-06 B20438-14 B20438-06
AAB20438-14EA 138.75 USD
CAAAB20438-06 AAB20438-14 CAAAB20438-14 AAB20438-06
2,4,4,6-Tetramethyl-1-oxa-3-aza-2-cyclohexene 95%
2,4,4,6-Tetramethyl-1-oxa-3-aza-2-cyclohexene
Formula: C₈H₁₅NO
Boiling Pt: 147…149 °C
Density: 0.886
Storage Temperature: Ambient
MDL Number: MFCD00014605
CAS Number: 26939-18-4
EINECS: 248-117-8
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR