Methyl-3-benzoylpropionate 98%

Synonyms: 3-Benzoylpropionic Acid Methyl EsterMethyl 4-Oxo-4-phenylbutyrate4-Oxo-4-phenylbutyric Acid Methyl Ester

B20435-06 B20435-14 B20435-06 B20435-14
AAB20435-06EA 78.28 USD
CAAAB20435-06 CAAAB20435-14 AAB20435-06 AAB20435-14
Methyl-3-benzoylpropionate 98%
Methyl-3-benzoylpropionate
Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Boiling Pt: 172…174 °C/10 mm
Storage Temperature: Ambient
MDL Number: MFCD00014906
CAS Number: 25333-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR