1-(5-Fluoro-2-methylphenyl)-2-thiourea 97%

Danger

Synonyms: N-(5-Fluoro-2-methylphenyl)thiourea

B20423-06 B20423-03 B20423-03 B20423-06
AAB20423-06EA 97.23 USD
CAAAB20423-03 AAB20423-06 CAAAB20423-06 AAB20423-03
1-(5-Fluoro-2-methylphenyl)-2-thiourea 97%
1-(5-Fluoro-2-methylphenyl)-2-thiourea
Formula: C₈H₉FN₂S
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00052380
CAS Number: 16822-86-9
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR