2,4-Dimethyl-3-heptanone 99%

Supplier: Thermo Scientific Chemicals
Warning

B20384-09 B20384-09 B20384-18 B20384-18
AAB20384-09EA 53.49 USD
AAB20384-09 CAAAB20384-09 AAB20384-18 CAAAB20384-18
2,4-Dimethyl-3-heptanone 99%
2,4-Dimethyl-3-heptanone
Formula: C₉H₁₈O
Boiling Pt: 170 °C
Density: 0.812
Flash Pt: 44 °C(111 °F)
Storage Temperature: Ambient
MDL Number: MFCD00027146
CAS Number: 18641-71-9
UN: 1224
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR