1-(3-Chlorophenyl)biguanide hydrochloride 97%
|
Warning
|
B20355-06
B20355-03
B20355-03
B20355-06
AAB20355-03EA
36.7
USD
CAAAB20355-06
AAB20355-03
AAB20355-06
CAAAB20355-03
1-(3-Chlorophenyl)biguanide hydrochloride 97%
1-(3-Chlorophenyl)biguanide hydrochloride
Specific agonist for 5-HT receptor
Formula:
C₈H₁₁Cl₂N₅ Melting Pt: 191…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053019 CAS Number: 2113-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...