6,8-Dibromo-2-oxo-2H-chromene-3-carboxylic acid 97%

Warning

B20222-03
CAAAB20222-03
6,8-Dibromo-2-oxo-2H-chromene-3-carboxylic acid 97%
6,8-Dibromo-2-oxo-2H-chromene-3-carboxylic acid
Formula: C₁₀H₄Br₂O₄
Melting Pt: 221…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00173735
CAS Number: 3855-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR