2-Chloro-2-methylhexane 97%

Warning

B20183-14 B20183-14
AAB20183-14EA 188.82 USD
CAAAB20183-14 AAB20183-14
2-Chloro-2-methylhexane 97%
2-Chloro-2-methylhexane
Formula: C₇H₁₅Cl
Storage Temperature: Ambient
MDL Number: MFCD00060779
CAS Number: 4398-65-6
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR