3,5-Dimethyl-3-hexanol 98%

Warning

B20141-06 B20141-06 B20141-03 B20141-03 B20141-14 B20141-14
AAB20141-03EA 34.92 USD
AAB20141-03 AAB20141-14 AAB20141-06 CAAAB20141-03 CAAAB20141-14 CAAAB20141-06
3,5-Dimethyl-3-hexanol 98%
3,5-Dimethyl-3-hexanol
Formula: C₈H₁₈O
Density: 0,83
Storage Temperature: Ambient
MDL Number: MFCD00021823
CAS Number: 4209-91-0
EINECS: 224-136-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR