4-Acetyldiphenyl sulfide 97%

Synonyms: 1-(4-(Phenylthio)phenyl)ethanone

B20073-06 B20073-03 B20073-03 B20073-06
AAB20073-03EA 57.56 USD
CAAAB20073-06 CAAAB20073-03 AAB20073-03 AAB20073-06
4-Acetyldiphenyl sulfide 97%
4-Acetyldiphenyl sulphide
Formula: C₁₄H₁₂OS
MW: 228.32 g/mol
Boiling Pt: 183…184 °C/3 mm
Melting Pt: 64…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00026227
CAS Number: 10169-55-8
EINECS: 233-443-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR