4-Bromo-3-methyl-1H-pyrazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-3-methylpyrazole

B20069-06 B20069-06 B20069-03 B20069-03
AAB20069-03EA 47.51 USD
AAB20069-03 CAAAB20069-06 CAAAB20069-03 AAB20069-06
4-Bromo-3-methyl-1H-pyrazole 97%
4-Bromo-3-methyl-1H-pyrazole
Formula: C₄H₅BrN₂
MW: 161.01 g/mol
Melting Pt: 73…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00005241
CAS Number: 13808-64-5
EINECS: 237-456-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR